Lanthanum(III) iodide, ultra dry ≥99.99%, powder
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Lanthanum triiodide
Ampuled under argon
047184-03
047184-06
AA47184-03EA
67.1
USD
AA47184-03
AA47184-06
Lanthanum(III) iodide, ultra dry ≥99.99%, powder
Lanthanum(III) iodide
Lanthanum(III) iodide, ultra dry, 99.99% (metals basis). Powder, Ampouled under argon.
Purity: 99.99%
CAS No.: 13813-22-4
Molecular Formula: LaI3
Formula Weight: 519.62
Physical Form: Powder, Ampouled under argon
Sensitivity: Moisture and Light Sensitive
Appearance: Off-white
Odor: Odorless
Melting Point: 772°
MDL No.: MFCD00049470
Formula:
I₃La MW: 519.62 g/mol Melting Pt: 778 Storage Temperature: Ambient |
MDL Number:
MFCD00049470 CAS Number: 13813-22-4 EINECS: 237-465-6 |
Specification Test Results
Assay Minimum | 99.99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...