Holmium(III) chloride, ultra dry ≥99.95%, beads

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Holmium trichloride

Ampuled under argon

047178-06 047178-14
AA47178-06EA 238 USD
AA47178-06 AA47178-14
Holmium(III) chloride, ultra dry ≥99.95%, beads
Holmium(III) chloride

Holmium(III) chloride, ultra dry, 99.95% (metals basis). -10 Mesh Beads, Ampouled under argon.


Purity: 99.95% CAS No.: 10138-62-2 Molecular Formula: HoCl3 Formula Weight: 271.29 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 3.70 Sensitivity: Hygroscopic Odor: Odorless Melting Point: 718° MDL No.: MFCD00011051

Formula: Cl₃Ho
MW: 271.29
Boiling Pt: 1500
Melting Pt: 718 °C
Density: 3.70
Storage Temperature: Ambient
MDL Number: MFCD00011051
CAS Number: 10138-62-2
EINECS: 233-387-1
Merck Index: 14,4727

Order Now

Specification Test Results

Assay Minimum 99.95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR