Bismuth(III) bromide, anhydrous ≥99.999% (metals basis), powder
Supplier: Thermo Scientific Chemicals
|
Danger
|
047175-03
047175-06
AA47175-03EA
97.18
USD
AA47175-03
AA47175-06
Bismuth(III) bromide, anhydrous ≥99.999% (metals basis), powder
Bismuth(III) bromide
Bismuth(III) bromide, anhydrous, 99.999% (metals basis). Powder.
Purity: 99.999%
CAS No.: 7787-58-8
Molecular Formula: BiBr3
Formula Weight: 448.71
Physical Form: Powder
Density: 5.72
Sensitivity: Hygroscopic
Appearance: Yellow
Odor: Acidic
Melting Point: 218°
Boiling Point: 453°
UN No.: 3260
MDL No.: MFCD00014195
Formula:
BiBr₃ MW: 448.69 g/mol Boiling Pt: 453 °C Melting Pt: 218 °C Density: 5.72 Storage Temperature: Ambient |
MDL Number:
MFCD00014195 CAS Number: 7787-58-8 EINECS: 232-121-1 UN: 3260 ADR: 8,III |
Specification Test Results
Assay Minimum | 99.999 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...