Aluminium bromide, ultra dry ≥99.999%, powder
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aluminum bromide
Ampuled under argon
047168-06
047168-09
AA47168-06EA
337
USD
AA47168-06
AA47168-09
Aluminium bromide, ultra dry ≥99.999%, powder
Aluminium bromide
Aluminum bromide, ultra dry, 99.999% (metals basis). Powder, Ampouled under argon.
Purity: 99.999%
CAS No.: 7727-15-3
Molecular Formula: AlBr3
Formula Weight: 266.71
Physical Form: Powder, Ampouled under argon
Density: 3.21
Sensitivity: Moisture Sensitive
Appearance: White to orange
Odor: Pungent
Melting Point: 97.5°
Boiling Point: 265°
UN No.: 1725
MDL No.: MFCD00003421
Formula:
AlBr₃ MW: 266.69 g/mol Boiling Pt: 265 Melting Pt: 97 Density: 3.21 Storage Temperature: Ambient |
MDL Number:
MFCD00003421 CAS Number: 7727-15-3 EINECS: 231-779-7 UN: 1725 ADR: 8 II Merck Index: 14,332 |
Specification Test Results
Assay Minimum | 99.999 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...