Manganese(II) iodide, ultra dry ≥99.99% (metals basis), beads -10 mesh

Supplier: Thermo Scientific Chemicals
Warning


ampouled under argon

047166-03 047166-06 047166-09
AA47166-03EA 103 USD
AA47166-03 AA47166-06 AA47166-09
Manganese(II) iodide, ultra dry ≥99.99% (metals basis), beads -10 mesh
Manganese(II) iodide

Manganese(II) iodide, ultra dry, 99.99% (metals basis). -10 Mesh Beads, Ampouled under argon.


Purity: 99.99% CAS No.: 7790-33-2 Molecular Formula: MnI2 Formula Weight: 308.75 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 5.0 Sensitivity: Light, Air and Moisture Sensitive Appearance: White to pink to red Odor: Odorless Melting Point: 638° MDL No.: MFCD00016225

Formula: I₂Mn
Melting Pt: 638 °C
Density: 5.0
Storage Temperature: Ambient
MDL Number: MFCD00016225
CAS Number: 7790-33-2
EINECS: 232-201-6
Merck Index: 14,5732

Order Now

Specification Test Results

Assay Minimum 99.99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR