Gallium triiodide, ultra dry ≥99.999% (metals basis), beads -10 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
ampouled under argon
047148-03
047148-06
047148-14
AA47148-03EA
82.9
USD
AA47148-03
AA47148-06
AA47148-14
Gallium triiodide, ultra dry ≥99.999% (metals basis), beads -10 mesh
Gallium triiodide
Gallium(III) iodide, ultra dry, 99.999% (metals basis). -10 Mesh Beads, Ampouled under argon.
Purity: 99.999%
CAS No.: 13450-91-4
Molecular Formula: GaI3
Formula Weight: 450.43
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 4.15
Sensitivity: Moisture and Light Sensitive
Appearance: Yellow-green
Melting Point: 212°
Boiling Point: 345° subl.
UN No.: 3260
MDL No.: MFCD00016107
Formula:
GaI₃ MW: 450.44 g/mol Boiling Pt: 345 °C (subl.) Melting Pt: 212 °C Density: 4.15 Storage Temperature: Ambient |
MDL Number:
MFCD00016107 CAS Number: 13450-91-4 EINECS: 236-611-6 UN: 3260 ADR: 8,II |
Specification Test Results
Assay Minimum | 99.999 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...