Dysprosium(III) bromide, ultra dry ≥99.99% (metals basis), beads -10 mesh
Supplier: Thermo Scientific Chemicals
|
Warning
|
ampouled under argon
047127-03
047127-06
AA47127-03EA
130
USD
AA47127-03
AA47127-06
Dysprosium(III) bromide, ultra dry ≥99.99% (metals basis), beads -10 mesh
Dysprosium(III) bromide
Dysprosium(III) bromide, ultra dry, 99.99% (metals basis). -10 Mesh Beads, Ampouled under argon.
Purity: 99.99%
CAS No.: 14456-48-5
Molecular Formula: DyBr3
Formula Weight: 402.23
Physical Form: -10 Mesh Beads, Ampouled under argon
Sensitivity: Hygroscopic
Melting Point: 881°
Boiling Point: 1480°
MDL No.: MFCD00016069
Formula:
Br₃Dy MW: 402.21 g/mol Boiling Pt: 1480 °C Melting Pt: 881 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016069 CAS Number: 14456-48-5 EINECS: 238-443-9 |
Specification Test Results
Assay Minimum | 99.99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...