Calcium iodide, ultra dry ≥99.995% (metals basis), beads -10 mesh

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Calcium iodide(Granules or Chunks)

ampouled under argon

047121-14 047121-22
AA47121-14EA 188 USD
AA47121-14 AA47121-22
Calcium iodide, ultra dry ≥99.995% (metals basis), beads -10 mesh
Calcium iodide

Calcium iodide, ultra dry, 99.995% (metals basis). -10 Mesh Beads, Ampouled under argon.


Purity: 99.995% CAS No.: 10102-68-8 Molecular Formula: CaI2 Formula Weight: 293.89 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 3.956 Sensitivity: Light Sensitive; Hygroscopic Appearance: White to pale yellow Odor: Odorless Melting Point: 784° Boiling Point: 1100° MDL No.: MFCD00010910

Formula: CaI₂
MW: 293.89 g/mol
Boiling Pt: 1100 °C
Melting Pt: 784 °C
Density: 3.956
Storage Temperature: Ambient
MDL Number: MFCD00010910
CAS Number: 10102-68-8
EINECS: 233-276-8
Merck Index: 14,1677

Order Now

Specification Test Results

Assay Minimum 99.995

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR