Indium(III) iodide, ultra dry ≥99.999%, beads -10 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
Ampuled under argon
047108-04
047108-09
047108-18
AA47108-04EA
63.5
USD
AA47108-04
AA47108-09
AA47108-18
Indium(III) iodide, ultra dry ≥99.999%, beads -10 mesh
Indium(III) iodide
Indium(III) iodide, ultra dry, 99.999% (metals basis). -10 Mesh Beads, Ampouled under argon.
Purity: 99.999%
CAS No.: 13510-35-5
Molecular Formula: InI3
Formula Weight: 495.53
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 4.69
Sensitivity: Light Sensitive; Hygroscopic
Appearance: Yellow to brown
Melting Point: 210°
UN No.: 3260
MDL No.: MFCD00016151
Formula:
I₃In MW: 495.53 g/mol Boiling Pt: 500 Melting Pt: 209.85 Density: 4.69 Storage Temperature: Ambient |
MDL Number:
MFCD00016151 CAS Number: 13510-35-5 EINECS: 236-839-6 UN: 3260 ADR: 8 III |
Specification Test Results
Assay Minimum | 99.999 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...