(2-Bromoethoxy)-tert-butyldimethylsilane 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Bromoethoxy)(tert-butyl)dimethylsilane

Stabilization: Stabilised with sodium carbonate (Na₂CO₃)

H56594-03 H56594-06
AAH56594-03EA 29.7 USD
AAH56594-03 AAH56594-06
(2-Bromoethoxy)-tert-butyldimethylsilane 98% stabilized
(2-Bromoethoxy)-tert-butyldimethylsilane

(2-Bromoethoxy)-tert-butyldimethylsilane, 98%


Purity: 98% CAS No.: 86864-60-0 Molecular Formula: C8H19BrOSi Formula Weight: 239.23 Flash Point: 73°(163°F) Physical Form: Liquid Density: 1.115 Refractive Index: 1.444 Appearance: Colorless Boiling Point: 120°/100mm MDL No.: MFCD00209550

Formula: C₈H₁₉BrOSi
MW: 239.228 g/mol
Boiling Pt: 120 °C/100 mm
Density: 1.115
Flash Pt: 73 °C(163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00209550
CAS Number: 86864-60-0

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR