Triphenyl borate 97%

Supplier: Thermo Scientific Chemicals
Danger

H55686-06 H55686-14
AAH55686-06EA 27.6 USD
AAH55686-06 AAH55686-14
Triphenyl borate 97%
Triphenyl borate

Triphenyl borate, 97%


Purity: 97% CAS No.: 1095-03-0 Molecular Formula: C18H15BO3 Formula Weight: 290.13 Physical Form: Solid Sensitivity: Moisture Sensitive Melting Point: 98-101° UN No.: 2811 MDL No.: MFCD00059011

Formula: C₁₈H₁₅BO₃
Melting Pt: 98…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00059011
CAS Number: 1095-03-0
EINECS: 214-137-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR