Tributylphenyltin 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Tri-n-butylphenyltin, 97%
Purity: 97%
CAS No.: 960-16-7
Molecular Formula: C18H32Sn
Formula Weight: 367.14
Flash Point: 113°(235°F)
Physical Form: Liquid
Density: 1.125
Refractive Index: 1.5160
Appearance: Colorless
Boiling Point: 125-128° /0.14mm
Beilstein Reference: 3610571
UN No.: 2788
MDL No.: MFCD00134394
Formula:
C₁₈H₃₂Sn Boiling Pt: 125…128° /0.14 mm Density: 1.125 Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00134394 CAS Number: 960-16-7 UN: 2788 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...