Methyl methanesulfonate 99%

Supplier: Thermo Scientific Chemicals
Danger

H55120-06 H55120-14 H55120-22
AAH55120-06EA 70.7 USD
AAH55120-06 AAH55120-14 AAH55120-22
Methyl methanesulfonate 99%
Methyl methanesulphonate

Methyl methanesulfonate, 99%


Purity: 99% CAS No.: 66-27-3 Molecular Formula: C2H6O3S Formula Weight: 110.13 Flash Point: 104°(219°F) Physical Form: Liquid Density: 1.3 Refractive Index: 1.4140 Appearance: Colorless to pale yellow Boiling Point: 202-203° Beilstein Reference: 1098586 Merck Reference: 14,6097 UN No.: 2810 MDL No.: MFCD00007557

Formula: C₂H₆O₃S
MW: 110.13 g/mol
Boiling Pt: 202…203 °C
Melting Pt: 20 °C
Density: 1.3 g/cm³ (20 °C)
Flash Pt: 104 °C (219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007557
CAS Number: 66-27-3
EINECS: 200-625-0
UN: 2810
ADR: 6.1,III
Merck Index: 12,06170

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR