1,3-Dibromoacetone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
1,3-Dibromoacetone, 98%
Purity: 98%
CAS No.: 816-39-7
Molecular Formula: C3H4Br2O
Formula Weight: 215.87
Storage Temperature: 2°C to 8°C
Flash Point: 102°(216°F)
Physical Form: Crystals or fused solid
Sensitivity: Moisture Sensitive
Appearance: White to pale yellow
Melting Point: 29-30°
Boiling Point: 97-98°/21mm
Beilstein Reference: 1740389
UN No.: 1759
MDL No.: MFCD00013540
Formula:
BrH₂CC(O)CH₂Br Boiling Pt: 97…98 °C/21 mm Melting Pt: 29…30 °C Flash Pt: 102 °C (216 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013540 CAS Number: 816-39-7 EINECS: 212-430-8 UN: 1759 ADR: 8,II |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...