N,N'-Bis(acryloyl)cystamine 98% for electrophoresis

Supplier: Thermo Scientific Chemicals
Warning


Packaged under argon.

J66893-03 J66893-06
AAJ66893-03EA 77.1 USD
AAJ66893-03 AAJ66893-06 AAJ66893-14
N,N'-Bis(acryloyl)cystamine 98% for electrophoresis
N,N'-Bis(acryloyl)cystamine

N,N'-Bis(acryloyl)cystamine, Electrophoresis Reagent, 98%. Packaged under argon.


Purity: 98% CAS No.: 60984-57-8 Molecular Formula: C10H16N2O2S2 Formula Weight: 260.37 Storage Temperature: -30°C to -10°C Physical Form: Powder, Packaged under argon Appearance: White Melting Point: 123-124° Solubility: Soluble at 50 mg/ml in acetic acid MDL No.: MFCD00036225

Formula: C₁₀H₁₆N₂O₂S₂
Melting Pt: 123…124 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036225
CAS Number: 60984-57-8
EINECS: 262-546-8

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR