Octyl-β-D-thioglucopyranoside 98+%, crystalline powder for electrophoresis
Supplier: Thermo Scientific Chemicals
n-Octyl-^b-D-thioglucopyranoside, Electrophoresis Grade, 98+%. Crystalline powder.
Purity: 98+%
CAS No.: 85618-21-9
Molecular Formula: C14H28O5S
Formula Weight: 308.44
Physical Form: Crystalline powder
Appearance: White
Melting Point: 128°
Beilstein Reference: 4182783
MDL No.: MFCD00012189
Formula:
C₁₄H₂₈O₅S Melting Pt: 128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012189 CAS Number: 85618-21-9 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...