3-Isobutyl-1-methylxanthine ≥99% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
2,6-Dihydroxy-3-isobutyl-1-methylpurine, IBMX, 3,7-Dihydro-3-isobutyl-1-methyl-1H-purine-2,6-dione, 3-Isobutyl-1-methyl-3,7-dihydro-1H-purine-2,6-dione
BML-PD140-1000
BML-PD140-0200
89161-340EA
330.27
USD
89161-340
76203-478
3-Isobutyl-1-methylxanthine ≥99% (by HPLC)
3-Isobutyl-1-methylxanthine
Potent, non-specific inhibitor of phosphodiesterases (IC50=2-50µM)
More potent than theophylline at adenosine receptors. Accelerates conversion of mouse fibroblast cells into adipose cells.
Formula:
C₁₀H₁₄N₄O₂ MW: 222.25 g/mol |
MDL Number:
MFCD00005584 CAS Number: 28822-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...