PGF2α High Sensitivity ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
Most sensitive PGF2α ELISA kit available for prostaglandin research.
For the quantitative determination of PGF2α in culture supernatants, milk, plasma, serum, saliva, and urine from any species.
The PGF2α high sensitivity EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 3 hours. Absorbance is read at 405 nm. Simply the most sensitive quantification fo PGF2α commercially available in an enzyme immunoassay. Assay more samples without extraction.
Información para pedidos: Contains DxS IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.