SCREEN-WELL® Adrenergic Ligand Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-2811-0500 BML-2811-0100
89156-268EA 2554.96 USD
89156-268 76002-246
SCREEN-WELL® Adrenergic Ligand Library, Enzo Life Sciences
Cell Culture Supplements and Additives
The adrenergic ligand library contains 84 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs
The library is ideal for screening or identifying recombinant orphan G protein-coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The adrenergic ligand library is 1 plate of the larger neurotransmitter library which contains over 650 CNS receptor ligands.