IP ActivFlo Biopsy III Premium Mesh Biopsy Cassettes, Leica
Supplier: Leica
39LC-625-8-L 39LC-625-10-L 39LC-625-4-L 39LC-625-9-L 39VWR-625-5-L 39LC-625-1-L 39LC-625-2-L 39LC-625-11-L 39LC-625-6-L
95041-522CS 402.6 USD
95041-522 95041-524 95041-512 95041-526 95041-514 95041-506 95041-516 95041-508 95041-520
IP ActivFlo Biopsy III Premium Mesh Biopsy Cassettes, Leica
Embedding Cassettes Biopsy Processing Cassettes
IP ActivFlo Biopsy III cassettes feature a patented “Fluidic Flow" pore design for maximum reagent exchange.
  • Suitable for securing the smallest specimens
  • Sloped walls
  • Available in a wide variety of colors
With a 0.26 mm pore size, the cassettes can secure the smallest specimens, eliminating the need for biopsy bags and sponges.
The cassettes also feature sloped walls, creating a basket for small tissue management. These two piece cassettes with hinged lid have been designed for the Leica IP C Cassette Printer, with <1% printing error rate.
Información para pedidos: Cassettes are available with taped packaging for easy printer loading.