Manzanita Wood Gnawing Sticks, Bio-Serv
Supplier: Bio-Serv
W0015 W0014 W0016
89412-154EA 11.8 USD
89412-154 89412-156 76353-800
Manzanita Wood Gnawing Sticks, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Made of natural manzanita hardwood, these gnawing sticks are extremely durable and long lasting
The unique color and interesting surface offer stimulating enrichment. Cagewasher safe and autoclavable.