Turf Foraging Crumbles™ Treats, Bio-Serv

Supplier: Bio-Serv
F05433 F05437
89067-408EA 117.23 USD
89067-408 89067-410
Turf Foraging Crumbles™ Treats, Bio-Serv
Animal Supplies Animal Feed
Small, fruit-flavored bits, available in a variety of flavors, are nutritionally complete foraging treats

  • Specially sized for use with Bio-Serv's Turf Foraging Board
  • Scatter into bedding to encourage rooting behavior in swine
  • Saves labor and less expensive than perishable fruit
  • Uncolored Banana and Pina Colada Flavors are Certified (Contaminant Screened) and suitable for use in GLP and TOX studies
  • Nutritionally assayed
  • Shelf Life: 12 months, cool dry storage

Ingredients: Dextrose, Casein, Cellulose, Corn Starch, Sucrose, Mineral Mix, Canola Oil, Maltodextrin, Tablet Binder, Choline Dihydrogen Citrate, Vitamin Mix, Ascorbic Acid, DL-Methionine, BHA, Natural and Artificial Flavors, Acesulfame Potassium, Citric Acid, Food Dye.

Caloric value: 3.34 kcal/g
Nutritional ash content: 4.6%
Nutritional carbohydrate content: 53.5%
Nutritional fat content: 5.1%
Nutritional fibre content: 11.8%
Nutritional moisture content: <10%
Nutritional protein content: 18.5%
Vitamin C: 4.5 mg/g
Vitamin D3: 2.1 mg/g
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR