Medi-Vac® Guardian™ Suction Canister Systems, Cardinal Health
Supplier: Cardinal Health
65652-531 65651-230 65651-220 65651-395 65651-515 65651-530 65651-212
59000-780CS 81.04 USD
59000-780 PH65651-230 PH65651-220 PH65651-395 PH65651-515 PH65651-530 PH65651-212
Medi-Vac® Guardian™ Suction Canister Systems, Cardinal Health
Safety Cans
The Guardian suction system features disposable hard, clear plastic containers
The Guardian LVC (large volume collection canister) tough, shatter-resistant hard plastic canisters feature sturdy, ergonomic handle and graduations for viewing and measuring contents. Available with two sizes of roll stands for easy mobility.
Our single-use Flex Advantage® liners are designed with safety features to provide maximum protection and ease of use during fluid collection, retention and disposal. Liners include a one-way valve built into the inner lid at the patient port to prevent backflow ofMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable