N-Ethyl-p-toluenesulfonamide, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
N-Ethyl-4-methylbenzenesulphonamide, p-Tolueneethylsulphonamide, N-Ethyltoluene-4-sulphonamide, N-Tosylethylamine
M-Clarity™ quality level = MQ200
8.43970.0050
8.43970.0250
EM8.43970.0050CS
1982
USD
EM8.43970.0050
EM8.43970.0250
N-Ethyl-p-toluenesulfonamide, Sigma-Aldrich®
N-Ethyl-p-toluenesulfonamide
N-Ethyltoluene-4-sulfonamide for synthesis
Formula:
C₉H₁₃NO₂S MW: 199.27 g/mol |
MDL Number:
MFCD00048511 CAS Number: 80-39-7 |
Specification Test Results
Hill Formula | C₉H₁₃NO₂S |
Molar Mass | 199.27 g/mol |
Density | 1.26 g/cm³ |
Flash point | 127 °C |
Melting Point | 63 - 65 °C |
Storage class | 10 - 13 Other liquids and solids |
WGK | WGK 3 highly water endangering |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...