Aniline, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Aminobenzene, Phenylamine
M-Clarity™ quality level = MQ200
8.22256.0100
8.22256.1000
EM8.22256.0100CS
293
USD
EM8.22256.0100
EM8.22256.1000
EM8.22256.5000
Aniline, Sigma-Aldrich®
Aniline
Aniline for synthesis
Formula:
C₆H₅NH₂ MW: 93.13 g/mol Boiling Pt: 184 Melting Pt: -6.2 Flash Pt: 76 |
MDL Number:
MFCD00007629 CAS Number: 62-53-3 UN: 1547 ADR: 6.1 II Merck Index: 13,00661 |
|
Specification Test Results
Hill Formula | C₆H₇N |
Chemical formula | C₆H₅NH₂ |
Molar Mass | 93.13 g/mol |
Boiling point | 184 °C (1013 hPa) |
Density | 1.02 g/cm³ (20 °C) |
Explosion limit | 1.2 - 11 %(V) |
Flash point | 76 °C |
Ignition temperature | 540 °C |
Melting Point | -6.2 °C |
pH value | 8.8 (36 g/l, H₂O, 20 °C) |
Vapor pressure | 0.5 hPa (20 °C) |
Refractive Index | 1.5863 (20 °C, 589 nm) |
Solubility | 36 g/l (20 °C) |
LD 50 dermal | LD50 Rabbit 840 mg/kg |
Storage class | 6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials |
WGK | WGK 2 water endangering |
R Phrase | R 23/24/25-40-41-43-48/23/24/25-68-50 |
S Phrase | S 26-27-36/37/39-45-46-61-63 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...