Rapid PNGase F
Supplier: New England Biolabs (NEB)
P0710S
P0711S
102877-828EA
537
USD
102877-828
103218-942
Rapid PNGase F
Enzymes
Rapid PNGase F is an improved reagent that allows the complete and rapid deglycosylation of antibodies and fusion proteins in minutes
- Non-reducing format
All N-glycans are released rapidly and without bias, and are ready to be prepared for downstream chromatography or mass spectrometry analysis. Rapid PNGase F creates an optimized workflow which reduces processing time without compromising sensitivity or reproducibility.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)