1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 4-Bromo-2-fluoro-1-(trifluoromethoxy)benzene

M-Clarity™ quality level = MQ200

8.43874.0010 8.43874.0050
EM8.43874.0010CS 237807 USD
EM8.43874.0010 EM8.43874.0050
1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene for synthesis, Sigma-Aldrich®
1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene
Formula: C₇H₃BrF₄O
MW: 259.00 g/mol
MDL Number: MFCD01861132
CAS Number: 105529-58-6
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Solubility 0.0288 g/l (20 °C)
Molar Mass 259 g/mol
Boiling Point 171 °C (994.5 hPa)
Vapor Pressure 4 hPa (25 °C)
Flash Point 69 °C
Density 1.63 g/cm3 (20 °C)
pH (H2O) neutral
Ignition Point 630 °C
Hill Formula C7H3BrF4O

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR