1,2,4-Trihydroxybenzene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.43791.0005 8.43791.0025
EM8.43791.0005CS 184595 USD
EM8.43791.0005 EM8.43791.0025
1,2,4-Trihydroxybenzene for synthesis, Sigma-Aldrich®
1,2,4-Trihydroxybenzene
Formula: C₆H₆O₃
MDL Number: MFCD00002198
CAS Number: 533-73-3

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 137 - 140 °C
Molar Mass 126.11 g/mol
Density 1.45 g/cm3 (20 °C)
Hill Formula C6H6O3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR