1-Phenylethyl acetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.43785.0100 8.43785.0500
EM8.43785.0100CS 1379 USD
EM8.43785.0100 EM8.43785.0500
1-Phenylethyl acetate for synthesis, Sigma-Aldrich®
1-Phenylethyl acetate
Formula: C₁₀H₁₂O₂
MDL Number: MFCD00008858
CAS Number: 93-92-5

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point -60 °C
Molar Mass 164.21 g/mol
Boiling Point 212 °C (1013 hPa)
Flash Point 91 °C
Density 1.03 g/cm3 (20 °C)
Ignition Point 450 °C
Hill Formula C10H12O2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR