Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV) ≥75% in 2-propanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Diisopropoxytitanium(IV) Bis(acetylacetonate), Titanium (di-i-propoxide)bis(acetylacetonate), Titanium(IV) Diisopropoxide Bis(acetylacetonate), Titanium (IV) bis (acetylacetonate) diisopropoxide

M-Clarity™ quality level = MQ200

8.43742.0100 8.43742.0500
EM8.43742.0100CS 1019 USD
EM8.43742.0100 EM8.43742.0500
Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV) ≥75% in 2-propanol for synthesis, Sigma-Aldrich®
Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV)
Formula: C₁₆H₂₈O₆Ti
MW: 364.26 g/mol
MDL Number: MFCD00000031
CAS Number: 17927-72-9
UN: 1219
ADR: 3 II

Order Now

Specification Test Results

Solubility (20 °C) Hydrolysis
Flash Point 12 °C
Density 1.01 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR