4-Methoxystyrene for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.41398.0005
8.41398.0025
EM8.41398.0005CS
15120
USD
EM8.41398.0005
EM8.41398.0025
4-Methoxystyrene for synthesis, Sigma-Aldrich®
4-Methoxystyrene
Formula:
C₉H₁₀O |
MDL Number:
MFCD00008619 CAS Number: 637-69-4 |
Specification Test Results
Molar Mass | 134.18 g/mol |
Boiling Point | 204 - 205 °C |
Flash Point | 76 °C |
Density | 1.002 g/cm3 (20 °C) |
Synonyms | 4-Vinylanisole |
Hill Formula | C9H10O |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...