Isophthalonitrile for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.41355.0100
8.41355.0500
EM8.41355.0100CS
5780
USD
EM8.41355.0100
EM8.41355.0500
Isophthalonitrile for synthesis, Sigma-Aldrich®
Isophthalonitrile
Formula:
C₈H₄N₂ |
MDL Number:
MFCD00001795 CAS Number: 626-17-5 UN: 2811 ADR: 6.1,II |
Specification Test Results
Solubility | 0.7 g/l (20 °C) |
Melting Point | 162 - 163 °C |
Molar Mass | 128.13 g/mol |
Bulk Density | 500 kg/m3 |
Boiling Point | 288 °C (1013 hPa) |
Vapor Pressure | 0.014 hPa (20 °C) |
Density | 1.3 g/cm3 (20 °C) |
Synonyms | Benzene-1,3-dicarbonitrile, 1,3-Dicyanobenzene |
Hill Formula | C8H4N2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...