2-(Hydroxymethyl)benzyl alcohol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
o-Phenylenedimethanol, o-Xylene-α,α'-diol, 1,2-Benzenedimethanol
M-Clarity™ quality level = MQ200
8.41297.0001
8.41297.0010
EM8.41297.0001CS
45520
USD
EM8.41297.0001
EM8.41297.0010
2-(Hydroxymethyl)benzyl alcohol for synthesis, Sigma-Aldrich®
2-(Hydroxymethyl)benzyl alcohol
Formula:
C₈H₁₀O₂ MW: 138.17 g/mol |
MDL Number:
MFCD00004626 CAS Number: 612-14-6 |
Specification Test Results
Melting Point | 62 - 64 °C |
Molar Mass | 138.16 g/mol |
Synonyms | 1,2-Bishydroxymethylbenzene |
Hill Formula | C8H10O2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...