Butyrophenone for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.41778.0025
8.41778.0250
EM8.41778.0025CS
3975
USD
EM8.41778.0025
EM8.41778.0250
Butyrophenone for synthesis, Sigma-Aldrich®
Butyrophenone
Formula:
C₆H₅C(O)(CH₂)₂CH₃ |
MDL Number:
MFCD00009397 CAS Number: 495-40-9 |
|
Specification Test Results
Melting Point | 11 - 13 °C |
Molar Mass | 148.2 g/mol |
Boiling Point | 220 - 222 °C (1013 hPa) |
Flash Point | 88 °C |
Density | 0.988 g/cm3 |
Hill Formula | C10H12O |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...