Dodecyltrimethylammonium bromide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Dodecyl trimethyl ammonium bromide, (1-Dodecyl)trimethylammonium bromide

M-Clarity™ quality level = MQ200

8.41022.0025 8.41022.0100
EM8.41022.0025CS 10696 USD
EM8.41022.0025 EM8.41022.0100
Dodecyltrimethylammonium bromide for synthesis, Sigma-Aldrich®
Dodecyltrimethylammonium bromide
Formula: C₁₅H₃₄BrN
MW: 308.35 g/mol
MDL Number: MFCD00011767
CAS Number: 1119-94-4
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Melting Point 237 - 245 °C
Molar Mass 308.34 g/mol
Hill Formula C15H34BrN

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR