Butyltin hydroxide oxide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.40126.0010 8.40126.0100
EM8.40126.0010CS 3745 USD
EM8.40126.0010 EM8.40126.0100
Butyltin hydroxide oxide for synthesis, Sigma-Aldrich®
Butyltin hydroxide oxide
Formula: C₄H₁₀O₂Sn
MDL Number: MFCD00013927
CAS Number: 2273-43-0
UN: 3146
ADR: 6.1,III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Molar Mass 208.81 g/mol
Boiling Point >350 °C
Vapor Pressure 1.47 hPa (20 °C)
Synonyms Butylhydroxyoxo stannane
Hill Formula C4H10O2Sn


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR