Benzoic acid methyl ester for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Methyl benzoate

M-Clarity™ quality level = MQ200

8.22330.1000 8.22330.2500
EM8.22330.1000CS 579 USD
EM8.22330.1000 EM8.22330.2500
Benzoic acid methyl ester for synthesis, Sigma-Aldrich®
Benzoic acid methyl ester
Formula: C₆H₅COOCH₃
MDL Number: MFCD00008421
CAS Number: 93-58-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=287e16081e44f5525ff8b4d34f0a6fece44a3073&vid=2be9af3b90e55af477c374c091cd0888dcf60b96&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.157 g/l (30 °C)
Melting Point -12 °C
Molar Mass 136.15 g/mol
Boiling Point 199 °C (1013 hPa)
Vapor Pressure 0.36 hPa (20 °C)
Flash Point 82 °C
Explosion Limit 8.6 - 20 %(V)
Density 1.09 g/cm3 (20 °C)
Ignition Point 510 °C
Hill Formula C8H8O2
Chemical Formula C6H5COOCH3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR