Methylstyrene (mixture of isomers) stabilized for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC)
M-Clarity™ quality level = MQ200
8.22098.0100
8.22098.1000
EM8.22098.0100CS
697
USD
EM8.22098.0100
EM8.22098.1000
Methylstyrene (mixture of isomers) stabilized for synthesis, Sigma-Aldrich®
Methylstyrene (mixture of isomers)
Formula:
C₉H₁₀ MW: 118.18 g/mol |
MDL Number:
MFCD00168066 CAS Number: 25013-15-4 UN: 2618 ADR: 3,III |
Specification Test Results
Solubility | 0.09 g/l (20 °C) |
Melting Point | -77 °C |
Molar Mass | 118.18 g/mol |
Boiling Point | 168 °C |
Vapor Pressure | 1.47 hPa (20 °C) |
Flash Point | 52 °C |
Explosion Limit | 1.1 - 5.2 %(V) |
Density | 0.893 g/cm3 (25 °C) |
Ignition Point | 575 °C |
Synonyms | Vinyltoluene |
Hill Formula | C9H10 |
Chemical Formula | 3/4-(CH3)C6H4CH=CH2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...