Dehydroacetic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 3-Acetyl-6-methyl-2H-pyran-2,4(3H)-dione, 3-Acetyl-4-hydroxy-6-methyl-2H-pyran-2-one

M-Clarity™ quality level = MQ200

8.22011.0005 8.22011.0250 8.22011.1000
EM8.22011.0005CS 31961 USD
EM8.22011.0005 EM8.22011.0250 EM8.22011.1000
Dehydroacetic acid for synthesis, Sigma-Aldrich®
Dehydroacetic acid
Formula: C₈H₈O₄
MW: 168.15 g/mol
MDL Number: MFCD00066709
CAS Number: 520-45-6

Order Now

Specification Test Results

Solubility 2 g/l (20 °C)
Melting Point 109 - 111 °C
Molar Mass 168.14 g/mol
Bulk Density 700 kg/m3
Boiling Point 270 °C (1013 hPa)
Vapor Pressure 0.001 hPa (20 °C)
Flash Point 157 °C
pH 4 (2 g/l, H2O, 20 °C)
Ignition Point 336 °C
Synonyms 3-Acetyl-3,4-dihydro-6-methyl-2H-pyran-2,4-dione
Hill Formula C8H8O4

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR