2,4-Dimethylbenzaldehyde for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.21461.0010
8.21461.0100
EM8.21461.0010CS
33631
USD
EM8.21461.0010
EM8.21461.0100
2,4-Dimethylbenzaldehyde for synthesis, Sigma-Aldrich®
2,4-Dimethylbenzaldehyde
Formula:
C₉H₁₀O MW: 134.18 g/mol |
MDL Number:
MFCD00003340 CAS Number: 15764-16-6 |
Specification Test Results
Solubility | (20 °C) slightly soluble |
Melting Point | -9 °C |
Molar Mass | 134.18 g/mol |
Boiling Point | 215 - 216 °C |
Vapor Pressure | 1 hPa (20 °C) |
Flash Point | 88 °C |
Refractive Index | 1.5492 (20 °C, 589 nm) |
Density | 1.02 g/cm3 (20 °C) |
Hill Formula | C9H10O |
Chemical Formula | 2,4-(CH3)2C6H3CHO |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...