Trityl alcohol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Triphenylmethanol, Triphenylcarbinol
M-Clarity™ quality level = MQ200
8.21196.0050
8.21196.0250
EM8.21196.0050CS
7765
USD
EM8.21196.0050
EM8.21196.0250
Trityl alcohol for synthesis, Sigma-Aldrich®
Trityl alcohol
Formula:
C₁₉H₁₆O MW: 260.34 g/mol |
MDL Number:
MFCD00004445 CAS Number: 76-84-6 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 160 - 163 °C |
Molar Mass | 260.34 g/mol |
Boiling Point | 360 °C |
Synonyms | Triphenyl carbinol |
Hill Formula | C19H16O |
Chemical Formula | (C6H5)3COH |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...