Sulphathiazole for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 4-Amino-N-(thiazol-2-yl)benzenesulphonamide, N1-2-Thiazolylsulphanilamid

M-Clarity™ quality level = MQ200

8.21112.0005 8.21112.0250
EM8.21112.0005CS 38300 USD
EM8.21112.0005 EM8.21112.0250
Sulphathiazole for synthesis, Sigma-Aldrich®
Sulfathiazole
Formula: C₉H₉N₃O₂S₂
MDL Number: MFCD00005319
CAS Number: 72-14-0

Order Now

Specification Test Results

Solubility 0.5 g/l (25 °C)
Melting Point 199 - 202 °C
Molar Mass 255.3 g/mol
Synonyms Sulfathiazol
Hill Formula C9H9N3O2S2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR