Phosphoryl tribromide for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Phosphorus(V) oxybromide, Phosphorus (V) oxide bromide, Phosphoryl bromide, Phosphorus oxybromide
M-Clarity™ quality level = MQ200
8.21026.0025
8.21026.0100
8.21026.1000
EM8.21026.0025CS
13072
USD
EM8.21026.0025
EM8.21026.0100
EM8.21026.1000
Phosphoryl tribromide for synthesis, Sigma-Aldrich®
Phosphoryl tribromide
Formula:
Br₃OP |
MDL Number:
MFCD00036298 CAS Number: 7789-59-5 UN: 1939 ADR: 8,II |
Specification Test Results
Solubility | (20 °C) (decomposition) |
Melting Point | 48 - 56 °C |
Molar Mass | 286.68 g/mol |
Boiling Point | 192 °C (1013 hPa) decomposes |
Density | 2.82 g/cm3 |
Synonyms | Phosphorus oxide tribromide, Phosphorus oxybromide |
Hill Formula | Br3OP |
Chemical Formula | POBr3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...