3-Oxoglutaric acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Acetone-1,3-dicarboxylic acid, 3-Ketoglutaric acid, 3-Oxopentanedioic acid, 1,3-Acetonedicarboxylic acid, β-Ketoglutaric acid

M-Clarity™ quality level = MQ200

8.20940.0050 8.20940.0250
EM8.20940.0050CS 9257 USD
EM8.20940.0050 EM8.20940.0250
3-Oxoglutaric acid for synthesis, Sigma-Aldrich®
3-Oxoglutaric acid
Formula: C₅H₆O₅
MW: 146.1 g/mol
MDL Number: MFCD00002711
CAS Number: 542-05-2

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 146.10 g/mol
Bulk Density 400 kg/m3
Synonyms ß-Ketoglutaric acid, Acetone-1,3-dicarboxylic acid
Hill Formula C5H6O5
Chemical Formula HOOCCH2COCH2COOH

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR