2'-Nitroacetanilide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: N-(2-Nitrophenyl)acetamide

M-Clarity™ quality level = MQ200

8.20879.0025 8.20879.0100
EM8.20879.0025CS 6204 USD
EM8.20879.0025 EM8.20879.0100
2'-Nitroacetanilide for synthesis, Sigma-Aldrich®
2'-Nitroacetanilide
Formula: C₈H₈N₂O₃
MW: 180.16 g/mol
MDL Number: MFCD00016991
CAS Number: 552-32-9

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 90 - 93 °C
Molar Mass 180.16 g/mol
Hill Formula C8H8N2O3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR