(S)-(-)-Nicotine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: (S)-(-)-3-(N-Methyl-2-pyrrolidinyl)pyridine, (S)-(-)-3-(1-Methyl-2-pyrrolidinyl)pyridine, L(-)-Nicotine, L-Nicotine, Nicotine, (-)-Nicotine

M-Clarity™ quality level = MQ200

8.20877.0025 8.20877.0100 8.20877.1000
EM8.20877.0025CS 4998 USD
EM8.20877.0025 EM8.20877.0100 EM8.20877.1000
(S)-(-)-Nicotine for synthesis, Sigma-Aldrich®
(S)-(-)-Nicotine
Formula: C₁₀H₁₄N₂
MW: 162.23 g/mol
MDL Number: MFCD00006369
CAS Number: 54-11-5
UN: 1654
ADR: 6.1,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -79 °C
Molar Mass 162.23 g/mol
Boiling Point 247 °C (1013 hPa)
Vapor Pressure 0.05 hPa (25 °C)
Flash Point 101 °C
Explosion Limit 0.7 - 4 %(V)
Density 1.01 g/cm3 (20 °C)
pH 10.2 (8.1 g/l, H2O, 20 °C)
Ignition Point 240 °C
Synonyms 3-(1-Methyl-2-pyrrolidinyl)pyridine, (S)-(-)-Nicotine
Hill Formula C10H14N2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR