2,7-Dihydroxynaphthalene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2,7-Naphthalenediol, Naphthalene-2,7-diol

M-Clarity™ quality level = MQ200

8.20851.0100 8.20851.0250
EM8.20851.0100CS 688 USD
EM8.20851.0100 EM8.20851.0250
2,7-Dihydroxynaphthalene for synthesis, Sigma-Aldrich®
2,7-Dihydroxynaphthalene
Formula: C₁₀H₈O₂
MW: 160.17 g/mol
MDL Number: MFCD00004085
CAS Number: 582-17-2

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 184 - 187 °C
Molar Mass 160.17 g/mol
Bulk Density 600 - 700 kg/m3
Synonyms 2,7-Dihydroxynaphthalene
Hill Formula C10H8O2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR