Thioanisole for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Methyl phenyl sulphide, Phenyl methyl sulphide, (Methylthio)benzene
M-Clarity™ quality level = MQ200
8.20825.0100
8.20825.0500
8.20825.2500
EM8.20825.0100CS
1119
USD
EM8.20825.0100
EM8.20825.0500
EM8.20825.2500
Thioanisole for synthesis, Sigma-Aldrich®
Thioanisole
Formula:
C₇H₈S MW: 124.21 g/mol |
MDL Number:
MFCD00008559 CAS Number: 100-68-5 UN: 3082 ADR: 9,III |
Specification Test Results
Solubility | 0.5 g/l (25 °C) |
Melting Point | -15 °C |
Molar Mass | 124.21 g/mol |
Boiling Point | 188 °C (1013 hPa) |
Flash Point | 57 °C |
Refractive Index | 1.5842 (25 °C, 589 nm) |
Density | 1.05 g/cm3 (20 °C) |
Synonyms | Thioanisole |
Hill Formula | C7H8S |
Chemical Formula | CH3SC6H5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...