2-Methylpentane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1,1-Dimethylbutane, Isohexane

M-Clarity™ quality level = MQ200

8.20818.0100 8.20818.0500
EM8.20818.0100CS 1001 USD
EM8.20818.0100 EM8.20818.0500
2-Methylpentane for synthesis, Sigma-Aldrich®
2-Methylpentane
Formula: C₆H₁₄
MW: 86.18 g/mol
MDL Number: MFCD00009406
CAS Number: 107-83-5
UN: 1208
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ce61b1646b637970765dc4b16a997650aeccfd1&vid=71606860b34952f73fd29b7df735dd0916b1e93c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.14 g/l (25 °C)
Melting Point -154 °C
Molar Mass 86.18 g/mol
Boiling Point 60 °C (1013 hPa)
Vapor Pressure 227 hPa (20 °C)
Flash Point -40 °C
Refractive Index 1.3715 (20 °C, 589 nm)
Explosion Limit 1.2 - 7.4 %(V)
Density 0.65 g/cm3 (20 °C)
Ignition Point 300 °C
Synonyms iso-Hexane, 1-Dimethylbutane
Hill Formula C6H14
Chemical Formula CH3CH2CH2CH(CH3)2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR