1,1,3,3-Tetramethylguanidine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
N,N,N',N'-Tetramethylguanidine
M-Clarity™ quality level = MQ200
8.21897.0100
8.21897.0250
EM8.21897.0100CS
1356
USD
EM8.21897.0100
EM8.21897.0250
1,1,3,3-Tetramethylguanidine for synthesis, Sigma-Aldrich®
1,1,3,3-Tetramethylguanidine
Formula:
C₅H₁₃N₃ MW: 115.18 g/mol |
MDL Number:
MFCD00008323 CAS Number: 80-70-6 UN: 2920 ADR: 8 (3),II |
Specification Test Results
Solubility | (20 °C) soluble |
Molar Mass | 115.17 g/mol |
Boiling Point | 162 °C (1013 hPa) |
Vapor Pressure | 0.26 hPa (25 °C) |
Flash Point | 50 °C |
Refractive Index | 1.4659 (20 °C, 589 nm) |
Explosion Limit | 1.0 - 7.5 %(V) |
Density | 0.92 g/cm3 (20 °C) |
pH | 12.7 (10 g/l, H2O, 25 °C) |
Ignition Point | 350 °C |
Hill Formula | C5H13N3 |
Chemical Formula | [(CH3)2N]2C=NH |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...